ENA018107796 Enamine

5-(4-Methoxy-phenyl)-5-methyl-imidazolidine-2,4-dione

  • MDL number MFCD00068872
  • Cas Number : 22927-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04374

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04374
Availability details for
Quantity
  • Availability
  • Add to Cart