ENA018107800 Enamine

2-chloro-N-{5-[ethyl(phenyl)sulfamoyl]-2-methoxyphenyl}acetamide

  • MDL number MFCD03964640
  • Cas Number : 568543-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-112
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04381
Availability details for
Quantity
  • Availability
  • Add to Cart