ENA018107801 Enamine

2-chloro-N-[2-methoxy-5-(piperidine-1-sulfonyl)phenyl]acetamide

  • MDL number MFCD03964672
  • Cas Number : 568543-70-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-116
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-04382

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04382
Availability details for
Quantity
  • Availability
  • Add to Cart