ENA018107805 Enamine

4-methyl-5,12-dithia-3-azatricyclo[7.3.0.0,2,6]dodeca-1,3,6,8,10-pentaen-8-ol

  • MDL number MFCD00456559
  • Cas Number : 294668-48-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 207-208
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04386

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04386
Availability details for
Quantity
  • Availability
  • Add to Cart