ENA018107851 Enamine

methyl 2-(2-chloroacetamido)-4,5-dimethylthiophene-3-carboxylate

  • MDL number MFCD02342280
  • Cas Number : 448199-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04444
Availability details for
Quantity
  • Availability
  • Add to Cart