ENA018107852 Enamine

6-phenylthieno[2,3-d]pyrimidine-4-thiol

  • MDL number MFCD02943933
  • Cas Number : 315685-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04445

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04445
Availability details for
Quantity
  • Availability
  • Add to Cart