ENA018107855 Enamine

2-(2-chloroacetamido)-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide

  • MDL number MFCD02341112
  • Cas Number : 436090-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-04448

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04448
Availability details for
Quantity
  • Availability
  • Add to Cart