ENA018107859 Enamine

7,9-dichloro-2-(chloromethyl)-6-methyl-4H-pyrido[1,2-a]pyrimidin-4-one

  • MDL number MFCD03964833
  • Cas Number : 726160-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04452

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04452
Availability details for
Quantity
  • Availability
  • Add to Cart