ENA018107865 Enamine

2-bromo-1-[4-(difluoromethoxy)phenyl]propan-1-one

  • MDL number MFCD03964598
  • Cas Number : 83022-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04459

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04459
Availability details for
Quantity
  • Availability
  • Add to Cart