ENA018107868 Enamine

1-(2-Trifluoromethyl-benzenesulfonyl)-pyrrolidine-2-carboxylic acid

  • MDL number MFCD03964629
  • Cas Number : 1009665-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04462
Availability details for
Quantity
  • Availability
  • Add to Cart