ENA018107886 Enamine

4-(2-methoxy-5-methylphenyl)-4-oxobutanoic acid

  • MDL number MFCD00235258
  • Cas Number : 55007-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04480

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04480
Availability details for
Quantity
  • Availability
  • Add to Cart