ENA018107890 Enamine

3-Bromo-4-oxo-4-p-tolyl-butyric acid

  • MDL number MFCD03964625
  • Cas Number : 53515-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116...118
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04485

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04485
Availability details for
Quantity
  • Availability
  • Add to Cart