ENA018107896 Enamine

7,7,9-Trimethyl-1,3-diaza-spiro[4.5]decane-2,4-dione

  • MDL number MFCD03953984
  • Cas Number : 91216-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 273...275
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04498

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04498
Availability details for
Quantity
  • Availability
  • Add to Cart