ENA018107898 Enamine

4-(2-methylcyclohexyl)-5-(pyridin-3-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03953983
  • Cas Number : 721893-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-04500

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04500
Availability details for
Quantity
  • Availability
  • Add to Cart