ENA018107899 Enamine

1-(3-chlorophenyl)-2,5-dimethyl-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD02605041
  • Cas Number : 428467-92-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-04501

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04501
Availability details for
Quantity
  • Availability
  • Add to Cart