ENA018107901 Enamine

3-methyl-1-phenyl-6-(thiophen-2-yl)-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD03956422
  • Cas Number : 721916-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04503

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04503
Availability details for
Quantity
  • Availability
  • Add to Cart