ENA018107903 Enamine

3-methyl-6-(thiophen-2-yl)-4-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxylic acid

  • MDL number MFCD03953981
  • Cas Number : 721893-48-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04505

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04505
Availability details for
Quantity
  • Availability
  • Add to Cart