ENA018107911 Enamine

2,5-Dimethyl-1-m-tolyl-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD02212416
  • Cas Number : 423749-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 43...45
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04515

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04515
Availability details for
Quantity
  • Availability
  • Add to Cart