ENA018107916 Enamine

4-Chlorosulfonyl-benzoic acid ethyl ester

  • MDL number MFCD00121815
  • Cas Number : 10486-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49...51
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04521

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-04521
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart