ENA018107919 Enamine

5-(3-bromophenyl)-5-methylimidazolidine-2,4-dione

  • MDL number MFCD03953995
  • Cas Number : 6943-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04524

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04524
Availability details for
Quantity
  • Availability
  • Add to Cart