ENA018107940 Enamine

5-[(1H-1,3-benzodiazol-1-yl)methyl]-4-(2-chlorophenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03955544
  • Cas Number : 721903-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04548

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04548
Availability details for
Quantity
  • Availability
  • Add to Cart