ENA018107942 Enamine

6-methyl-3-sulfanyl-[1,2,4]triazolo[4,3-b][1,2,4]triazin-7-ol

  • MDL number MFCD03955542
  • Cas Number : 877-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-04550

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04550
Availability details for
Quantity
  • Availability
  • Add to Cart