ENA018107948 Enamine

2-[4-(phenylamino)phenoxy]acetic acid

  • MDL number MFCD03950789
  • Cas Number : 643753-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-131
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-04560

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04560
Availability details for
Quantity
  • Availability
  • Add to Cart