ENA018107957 Enamine

1-carbamoylpyrrolidine-2-carboxylic acid

  • MDL number MFCD03936200
  • Cas Number : 125411-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-04570

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04570
Availability details for
Quantity
  • Availability
  • Add to Cart