ENA018107960 Enamine

2-(ethanesulfonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

  • MDL number MFCD03964556
  • Cas Number : 1008987-50-5
  • Molecular Weight : 269.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-111
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04574

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04574
Availability details for
Quantity
  • Availability
  • Add to Cart