ENA018107961 Enamine

ethyl 2-(2-chloroacetamido)-4-(thiophen-2-yl)thiophene-3-carboxylate

  • MDL number MFCD01121929
  • Cas Number : 315676-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-04576

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04576
Availability details for
Quantity
  • Availability
  • Add to Cart