ENA018107970 Enamine

ethyl 2-(2-chloroacetamido)-5-(diethylcarbamoyl)-4-methylthiophene-3-carboxylate

  • MDL number MFCD01352304
  • Cas Number : 550351-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-04587

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04587
Availability details for
Quantity
  • Availability
  • Add to Cart