ENA018107971 Enamine

2-(5,6-Dimethyl-thieno[2,3-d]pyrimidin-4-ylsulfanyl)-benzoic acid

  • MDL number MFCD03964551
  • Cas Number : 726151-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200...202
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04588

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04588
Availability details for
Quantity
  • Availability
  • Add to Cart