ENA018107975 Enamine

1-[5-chloro-3-(chloromethyl)-2-hydroxyphenyl]ethan-1-one

  • MDL number MFCD03964580
  • Cas Number : 34987-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 48-49
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04592

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04592
Availability details for
Quantity
  • Availability
  • Add to Cart