ENA018107986 Enamine

2-[4-(4-methylphenyl)-2-(pyridin-3-yl)-1,3-thiazol-5-yl]acetic acid

  • MDL number MFCD03988003
  • Cas Number : 554438-65-2
  • Molecular Weight : 310.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-04604

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04604
Availability details for
Quantity
  • Availability
  • Add to Cart