ENA018107994 Enamine

ethyl 2-(2-cyanoacetamido)-4-(4-methylphenyl)thiophene-3-carboxylate

  • MDL number MFCD03393074
  • Cas Number : 546071-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04615

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04615
Availability details for
Quantity
  • Availability
  • Add to Cart