ENA018107998 Enamine

1-Mercapto-4-(4-methyl-benzyl)-4H-[1,2,4]triazolo[4,3-a]quinazolin-5-one

  • MDL number MFCD03956543
  • Cas Number : 722465-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04619

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04619
Availability details for
Quantity
  • Availability
  • Add to Cart