ENA018108001 Enamine

4-(2-fluorophenyl)-5-(pyridin-3-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03956545
  • Cas Number : 722465-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-04622

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04622
Availability details for
Quantity
  • Availability
  • Add to Cart