ENA018108012 Enamine

3-(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)-2-mercapto-3H-quinazolin-4-one

  • MDL number MFCD03956553
  • Cas Number : 119426-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04633

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04633
Availability details for
Quantity
  • Availability
  • Add to Cart