ENA018108013 Enamine

5-(2-chlorophenyl)-4-(4-methoxyphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03803820
  • Cas Number : 370848-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-04634

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04634
Availability details for
Quantity
  • Availability
  • Add to Cart