ENA018108024 Enamine

4-(4-Fluoro-phenyl)-4-oxo-but-2-enoic acid

  • MDL number MFCD01656421
  • Cas Number : 777-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -...
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04649

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04649
Availability details for
Quantity
  • Availability
  • Add to Cart