ENA018108043 Enamine

1-carbamoyl-4-hydroxypyrrolidine-2-carboxylic acid

  • MDL number MFCD03960611
  • Cas Number : 1098066-21-7
  • Molecular Weight : 174.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-04674

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04674
Availability details for
Quantity
  • Availability
  • Add to Cart