ENA018108044 Enamine

2-chloro-N-(3,5-dichloro-4-methylpyridin-2-yl)acetamide

  • MDL number MFCD03964546
  • Cas Number : 726150-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04675

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04675
Availability details for
Quantity
  • Availability
  • Add to Cart