ENA018108048 Enamine

2-{[5-(4-chlorophenyl)-1,3,4-oxadiazol-2-yl]sulfanyl}acetic acid

  • MDL number MFCD02168632
  • Cas Number : 49809-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-144
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-04680

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04680
Availability details for
Quantity
  • Availability
  • Add to Cart