ENA018108057 Enamine

2-{1-benzyl-3-methyl-2,4,6-trioxo-1H,2H,3H,4H,5H,6H,7H-pyrrolo[2,3-d]pyrimidin-5-yl}acetic acid

  • MDL number MFCD03964578
  • Cas Number : 726152-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 235-236
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04693

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04693
Availability details for
Quantity
  • Availability
  • Add to Cart