ENA018108072 Enamine

5-[(2-phenylphenoxy)methyl]-4-(prop-2-en-1-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03956504
  • Cas Number : 722464-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-04709

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04709
Availability details for
Quantity
  • Availability
  • Add to Cart