ENA018108080 Enamine

2-{[(furan-2-yl)methyl]amino}benzoic acid

  • MDL number MFCD01025564
  • Cas Number : 501661-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-131
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04721

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04721
Availability details for
Quantity
  • Availability
  • Add to Cart