ENA018108083 Enamine

2-{[2-(cyclohex-1-en-1-yl)ethyl]amino}benzoic acid

  • MDL number MFCD03956563
  • Cas Number : 722466-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94-95
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-04724

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04724
Availability details for
Quantity
  • Availability
  • Add to Cart