ENA018108097 Enamine

4-(3-chloro-4-methoxyphenyl)-5-phenyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03956464
  • Cas Number : 721916-33-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-04744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04744
Availability details for
Quantity
  • Availability
  • Add to Cart