ENA018108099 Enamine

4-(2-fluorophenyl)-5-(4-methoxyphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03956567
  • Cas Number : 722466-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210-211
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04746

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04746
Availability details for
Quantity
  • Availability
  • Add to Cart