ENA018108100 Enamine

2-amino-4-(pyrrolidine-1-sulfonyl)phenol

  • MDL number MFCD03964564
  • Cas Number : 726151-75-3
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-4-(1-pyrrolidinylsulfonyl)phenol
Inchi Code 1S/C10H14N2O3S/c11-9-7-8(3-4-10(9)13)16(14,15)12-5-1-2-6-12/h3-4,7,13H,1-2,5-6,11H2
Inchi Key BUWCJYVBLURXKW-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-04747

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-04747
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart