ENA018108115 Enamine

5-(4-Ethoxy-phenoxy)-3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD03960593
  • Cas Number : 882221-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04762

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04762
Availability details for
Quantity
  • Availability
  • Add to Cart