ENA018108117 Enamine

4-(4-tert-butylphenoxy)-3-nitrobenzaldehyde

  • MDL number MFCD03964557
  • Cas Number : 726151-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04764

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04764
Availability details for
Quantity
  • Availability
  • Add to Cart