ENA018108161 Enamine

4-(5-bromo-2-methoxyphenyl)-4-oxobutanoic acid

  • MDL number MFCD03988537
  • Cas Number : 568567-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04814

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04814
Availability details for
Quantity
  • Availability
  • Add to Cart