ENA018108164 Enamine

4-Oxo-4-(4-pentyloxy-phenyl)-but-2-enoic acid

  • MDL number MFCD02668226
  • Cas Number : 27959-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04817

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04817
Availability details for
Quantity
  • Availability
  • Add to Cart